This product is now discontinued, View Related Products
Disposable Capsule Filters, Sterile Nylon
These capsule filters are highly retentive and come in an all-polypropylene housing for excellent chemical compatibility and flow. Since no adhesives, binders, or surfactants are used in manufacturing, these capsules offer low filter extractables. The sterile nylon filter capsules are naturally hydrophilic and are specifically designed for liquid applications; no prewetting necessary.
$97.24 - $158.68CAD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Connections Membrane Material Pore Size (micron)
Availability Pricing
ZenPure Disposable Sterile Nylon Membrane Filter Capsule; 0.2um RK-49700-40 Mfr # MKLN020D2H2H-ETO
COMPARE
1/4\" to 3/8\" hose barb Nylon 0.2
$97.24CAD / Each
ZenPure Disposable Sterile Nylon Membrane Filter Capsule; 1.0um, 1/4-3/8" barb RK-49700-42 Mfr # MKLN120D2H2H-ETO
COMPARE
1/4\" to 3/8\" hose barb Nylon 1
$110.80CAD / Each
ZenPure Disposable Sterile Nylon Membrane Filter Capsule; 0.2um; 1-4-3/8" barb RK-49700-50 Mfr # SKLS020H2H2H-PH-ETO
COMPARE
1/4\" to 3/8\" hose barb Nylon 0.2
$156.33CAD / Each
ZenPure Disposable Sterile Nylon Membrane Filter Capsule; 0.45um, 1/4-3/8" barb RK-49700-52 Mfr # SKLN045H2H2H-ETO
COMPARE
1/4\" to 3/8\" hose barb Nylon 0.45
$156.33CAD / Each
ZenPure Disposable Sterile Nylon Membrane Filter Capsule; 1.0um, 1/2" barb RK-49700-54 Mfr # SKLN120H2H2H-ETO
COMPARE
1/2\" hose barb Nylon 1
$158.68CAD / Each