This product is now discontinued, View Related Products
DispoPipette Disposable Liquid Samplers
Ideal for sampling low viscosity fluids
  • Two way sampling—all layer and target point
  • Cleanroom manufactured
  • Individually wrapped
These single-use HDPE samplers feature two different size openings (2.5 and 5.5 mm) at each end to accommodate sampling of different viscosity liquids. Simply insert an open-top sampler into the liquid to take an all-layer sample; insert while closed then open to obtain a target-point sample. DispoPipette samplers are manufactured in a cleanroom and individually packaged to ensure accurate sampling results.
$218.06 - $303.22CAD / Pkg
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Length (in) Description
Availability Pricing
Dynalon DispoPipette Samplers, 50 cm L, 100 mL, HDPE; Pk of 20 RK-33687-71 Mfr # 853933-5500
COMPARE
19.625 DispoPipette Samplers, 50 cm L, 100 mL, HDPE; Pk of 20
$218.06CAD / Pkg
Dynalon DispoPipette Samplers, Sterile, 50 cm L, 100 mL, HDPE; Pk of 20 RK-33687-72 Mfr # 853933-5501
COMPARE
19.625 DispoPipette Samplers, Sterile, 50 cm L, 100 mL, HDPE; Pk of 20
$247.09CAD / Pkg
Dynalon DispoPipette Samplers, 100 cm L, 200 mL, HDPE; Pk of 20 RK-33687-73 Mfr # 853933-5531
COMPARE
39.375 DispoPipette Samplers, 100 cm L, 200 mL, HDPE; Pk of 20
$251.95CAD / Pkg
Dynalon DispoPipette Samplers, Sterile, 100 cm L, 200 mL, HDPE; Pk of 20 RK-33687-74 Mfr # 853933-5532
COMPARE
39.375 DispoPipette Samplers, Sterile, 100 cm L, 200 mL, HDPE; Pk of 20
$303.22CAD / Pkg