This product is now discontinued, View Related Products
Dilution Vials
Polystyrene vials have optically clear parallel sides and a square bottom. They have been deionized and destaticized, eliminating particle interference in optical cell counters. The polystyrene snap-cap is leakproof, but easy to remove. Vials are guaranteed to meet or exceed OEM specifications. Each lot is tested to ensure cleanliness. Available in vacuum formed trays or in bulk packaging.
$235.74 - $297.62CAD / Case of 1000
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Height (mm) Volume (mL)
Availability Pricing
Simport Dilution Vials in Trays, 1000/CS RK-08918-50 Mfr # V130
COMPARE
56 25
$297.62CAD / Case of 1000
Simport Dilution Vials Bulk Pack, 1000/CS RK-08918-55 Mfr # V130-1
COMPARE
56 25
$235.74CAD / Case of 1000