This product is now discontinued, View Related Products
Digital Vibration Meters
Precision vibration meters help eliminate down time and costly repairs
  • Measures overall vibration amplitude in the range of 60 to 420,000 RPMs
  • Industrial 100 mV/g Accelerometer
  • Accelerometer for measuring vibration displacement, velocity and acceleration
  • Lightweight, rugged design for industrial environments
  • Easy-to-read LCD Graphic Display
  • Magnetic Base
  • 7" Probe Extention
  • Auto power off
  • English and metric models
$1,436.56CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Digital Vibration Meter, English Version
$1,436.56CAD / Each
Balmac Inc. 235M Digital Vibration Meter, Metric Version RK-65700-21 Mfr # 235M
COMPARE
Digital Vibration Meter, Metric Version
$1,436.56CAD / Each