Cole-Parmer® Thermocouple Wire - 30 Gauge
Make your own extension cables
Wire comes in 30 gauge size, ideal for fabricating your own probes or extension cables (meets ANSI and ASTM standards). Choose from wire with fluorinated ethylene propylene resin (FEP/FEP) or fiberglass materials as well as in 100-ft or 1000-ft lengths for any application.
$137.03 - $1,245.01CAD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (ft) Material Max Temperature (° F)
Availability Pricing
COMPARE
100 Fluorinated ethylene propylene (FEP) 400
$160.60CAD / Each
COMPARE
100 Fiberglass 900
$137.03CAD / Each
COMPARE
100 Fluorinated ethylene propylene (FEP) 400
$181.23CAD / Each
COMPARE
100 Fiberglass 900
$147.34CAD / Each
COMPARE
100 Fluorinated ethylene propylene (FEP) 400
$184.17CAD / Each
COMPARE
1000 Fiberglass 900
$844.99CAD / Each
COMPARE
1000 Fluorinated ethylene propylene (FEP) 400
$1,228.81CAD / Each
COMPARE
1000 Fiberglass 900
$1,046.11CAD / Each
COMPARE
1000 Fluorinated ethylene propylene (FEP) 400
$1,245.01CAD / Each
COMPARE
1000 Fiberglass 900
$1,019.59CAD / Each
Showing 1- 10 of 10