This product is now discontinued, View Related Products
Digi-Sense Thermocouple Industrial Elements
Heavy-duty construction for high temperature use
Elements with in an industrial thermocouple that includes a thermowells that deteriorate over time can be replaced as a more cost effective solution. Each replacement element is made of 20 gauge wire and is encased with ceramic insulators for a greater degree of protection.
$28.73 - $36.10CAD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° F) Probe Type
Availability Pricing
COMPARE
2300 K
$28.73CAD / Each
COMPARE
2300 K
$28.73CAD / Each
COMPARE
2300 K
$32.78CAD / Each
COMPARE
2300 K
$36.10CAD / Each
COMPARE
2300 J
$28.73CAD / Each
COMPARE
2300 J
$28.73CAD / Each
COMPARE
2300 J
$32.78CAD / Each
COMPARE
2300 J
$36.10CAD / Each
Showing 1- 8 of 8