Digi-Sense Solar-Powered Adjustable-Angle Industrial Thermometers
View readings from almost any angle by simply adjusting the angle of the head. Three-inch diameter adjustable head has an easy-to-read LCD. Requiring no batteries these thermometers are ideal in pilot plant or research and development applications where exact readings are required. The safety circuit prevents false readings.
Each thermometer is housed in a weather-resistant 304 stainless steel (SS) case. Stem is 304 SS. An acrylic lens protects the face. To further protect thermometer order a thermowell from page 1052. Simply match the stem length to the probe depth "A" for the thermowell.
$442.39 - $451.59CAD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Max Temperature (° F) Overall Length (mm)
Availability Pricing
COMPARE
300
$442.39CAD / Each
COMPARE
300
$442.39CAD / Each
COMPARE
300
$442.39CAD / Each
COMPARE
150
$451.59CAD / Each
COMPARE
150
$442.39CAD / Each
COMPARE
150
$442.39CAD / Each