This product is now discontinued, View Related Products
Digi-Sense RTD Twin Temp Industrial Probes
Critical process data can be captured remotely while still allowing local monitoring by the operator
Cole Parmer Twin Temp thermometers are the perfect solution for systems requiring local and remote temperature readings. Local readings are provided by either an analog gauge. The remote reading is achieved by a continuous 4-20 mA output signal. All sensors are located in the thermometer stem and will easily fit into existing bi-metal thermowells of corresponding length. Housings are manufactured out of rugged 304 stainless guaranteed hermetically sealed to prevent fogging.
$531.89 - $825.10CAD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Connector Type Max Temperature (° F)
Availability Pricing
COMPARE
1/2\" NPT(M) 250
$801.52CAD / Each
COMPARE
1/2\" NPT(M) 250
$825.10CAD / Each
COMPARE
1/2\" NPT(M) 250
$825.10CAD / Each
COMPARE
1/2\" NPT(M) 250
$825.10CAD / Each
COMPARE
1/2\" NPT(M) 500
$825.10CAD / Each
COMPARE
1/2\" NPT(M) 500
$801.52CAD / Each
COMPARE
1/2\" NPT(M) 500
$531.89CAD / Each
COMPARE
1/2\" NPT(M) 500
$825.10CAD / Each
Showing 1- 8 of 8