This product is now discontinued, View Related Products
Digi-Sense RTD Jack Panels for FS Conduit Box
Connectors withstand continuous temperature of 350°F (177°C) and maximum temperature of 400°F (204°C)
Feature glass-filled nylon casing with finger grips and polarity indicators; negative terminals are marked with red discs for easy identification. Miniconnectors accept miniature cable clamps and cable up to 20 gauge; standard connectors accept standard cable clamps and cable up to 16 gauge.
$35.73 - $226.53CAD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Connector Type Material Use With Probe / Wire Type
Availability Pricing
COMPARE
Mini-Connector
$35.73CAD / Each
COMPARE
Mini-Connector
$74.41CAD / Each
COMPARE
Standard Connector
$66.74CAD / Each
COMPARE
Standard Connector
$76.25CAD / Each
COMPARE
Standard Connector
$136.66CAD / Each
COMPARE
Mini-Connector
$41.62CAD / Each
COMPARE
Mini-Connector
$65.57CAD / Each
COMPARE
Mini-Connector
$99.82CAD / Each
COMPARE
Standard Connector
$222.48CAD / Each
COMPARE
Standard Connector
$226.53CAD / Each
Showing 1- 10 of 10