Digi-Sense RTD Industrial Probes with Thermowell
All-in-one, includes industrial probe head, element and thermowell
Industrial RTDs assemblies produce greater accuracy are ideal for use in industrial applications, chemical processes, production lines, ovens, furnaces and vessels. . The probes conductor is made of 24 gauge cooper wire and is encased with FEP insulation, and has a 316 stainless steel sheath. Featuring a standard size protection head with a gasketed cap to seal out dust and moisture and houses a ceramic terminal block. Head has a 3/4" NPT (F) opening on the side for easy connection to electrical conduit. 316 stainless steel thermowells are included in assemble and are ideal for applications where removing of the probe without draining the vessel is necessary.
$405.18 - $584.20CAD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Sheath material
Availability Pricing
COMPARE
350
$408.13CAD / Each
COMPARE
350
$436.12CAD / Each
COMPARE
350
$505.37CAD / Each
COMPARE
350
$559.89CAD / Each
COMPARE
350
$405.18CAD / Each
COMPARE
350
$427.28CAD / Each
COMPARE
350
$584.20CAD / Each
Showing 1- 7 of 7