Digi-Sense Red Spirit Incubator Glass Thermometers
Replace mercury thermometers for a specific application with the same accuracy without the any hazards
  • Non-mercury liquid
These thermometers are designed for use in specific applications. These Durac thermometers feature markings that are permanently printed on the glass, individually serialized, and red spirit liquid that has a dark scale contrasts against a white background. Some thermometers are PFA coated for added safety which may cause a slight change in accuracy up to 1 division.
Incubator thermometers - designed with a swell in the glass to hold the thermometer in place.
$78.09CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Min Temperature (° C) Overall Length (mm)
Availability Pricing
COMPARE
60 18 122
$78.09CAD / Each
COMPARE
110 50 150
$78.09CAD / Each