Cole-Parmer® Irreversible High-Temperature 4-Point Micro Horizontal Labels / Indicators
Reliable proof that the product was maintained at the correct temperature
  • Used as a quick inspection tool that the product is in usable condition during the shipping and receiving process
  • Save time and money due to costly recalls
  • Self-adhesive design
Labels are made up of a series of temperature-sensitive elements sealed between heat-resistant substrates with transparent windows. Each element changes color distinctly as its rated temperature is met or exceeded. The changes to the label are irreversible, providing a temperature history of the surface being monitored. The labels will not delaminate when removed, allowing them to be attached to an inspection report to serve as a permanent record. Ideal for use in the food, automotive, molding and metal working industries. Accuracy is ±1.8°F below 212°F and ±1°F above 212°F. Shelf life: 1 year after date of purchase.
$45.09 - $48.62CAD / Pkg of 10
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Height (in) Max temperature (° F) Temperature Points (° F)
Availability Pricing
COMPARE
11/64 190 160, 171, 180, 190
$48.62CAD / Pkg of 10
COMPARE
11/64 309 280, 289, 300, 309
$45.09CAD / Pkg of 10
COMPARE
11/64 351 320, 331, 340, 351
$45.09CAD / Pkg of 10