This product is now discontinued, View Related Products
Digi-Sense Dampened Back-Connected Dual-Scale Industrial Bimetal Thermometers
The benefits of a liquid-filled case in a silicone-free dry configuration
  • Dampened movement
  • More economical than silicone gauges
  • Greater temperature range than a silicone-filled gauges
  • Anti-parallax dial for accurate readings
  • Eliminate the risk of a leaking case and cross-contamination
  • Hermetically sealed per ASME B40.3 standard to prevent fogging and protect against tough environments
  • Rugged 304 stainless steel construction with a clear glass lens
  • Recalibrate using the external reset screw
Accuracy is ±1% full-scale when stem is immersed 2" in liquids or 4" in gases (immerse models with a 2.5" stem length 2" in gases). Measured stem length from the top of the threads to the bottom tip of the stem. Stem dia is 0.25"; threaded connection is 1/2" NPT(M). All thermometers meet or exceed ASME B40.3 standards. Maximum environmental temperature for glass lens thermometers is 500°F (148°C).
$91.35 - $186.38CAD / Each
23 variations of this product are available.
See All Product Options Below
23 PRODUCT OPTIONS
Compareitem specifications
Dial Size (in) Max Temperature (° F) Stem Length (in)
Availability Pricing
COMPARE
3 250 2.5
$93.56CAD / Each
COMPARE
3 550 2.5
$140.71CAD / Each
COMPARE
3 550 4
$91.35CAD / Each
COMPARE
3 250 6
$93.56CAD / Each
COMPARE
3 550 6
$93.56CAD / Each
COMPARE
3 250 9
$93.56CAD / Each
COMPARE
3 550 9
$93.56CAD / Each
COMPARE
3 250 12
$103.14CAD / Each
COMPARE
3 550 12
$100.19CAD / Each
COMPARE
3 250 24
$140.71CAD / Each
COMPARE
3 550 24
$140.71CAD / Each
COMPARE
5 250 2.5
$127.08CAD / Each
COMPARE
5 550 2.5
$116.40CAD / Each
COMPARE
5 250 4
$125.97CAD / Each
COMPARE
5 550 4
$129.66CAD / Each
COMPARE
5 250 6
$127.08CAD / Each
COMPARE
5 550 6
$127.08CAD / Each
COMPARE
5 250 9
$127.08CAD / Each
COMPARE
5 550 9
$127.08CAD / Each
COMPARE
5 250 12
$130.40CAD / Each
Showing 1 - 20 of 23