Digi-Sense® Blue Spirit Laboratory Standard Accuracy Glass Thermometers, Farhenheit-scale
Scales are permanently fused on to the glass and won’t wear off
  • Nonmercury blue spirit liquid
  • Statement of accuracy
These thermometers are inexpensive and safe for use in laboratories, universities, food/beverage, environmental, wastewater, petrochemical, pharmaceutical and many other general-purpose applications. Choose from partial immersion or total immersion thermometers.
Thermometers feature permanently fused markings, blue spirit liquid, white background glass; are also individually serial numbered. Include a ring top and a statement of accuracy supplied by manufacturer.
$31.24 - $40.67CAD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Immersion Depth (mm) Max Temperature (° F) Min Temperature (° F)
Availability Pricing
COMPARE
76 120 -40
$39.19CAD / Each
COMPARE
76 300 0
$31.24CAD / Each
COMPARE
76 500 20
$40.67CAD / Each
COMPARE
300 120 -40
$39.19CAD / Each
COMPARE
300 230 0
$31.68CAD / Each
COMPARE
300 300 0
$34.62CAD / Each