This product is now discontinued, View Related Products
Digi-Sense AM1950 and AM1960 Series Standard Platinum Resistance Thermometers SPRT Probes with Quartz Sheath
State-of-the-art, high-accuracy temperature probes at a fraction of the cost
  • Equivalent to Fluke Calibration / Hart Scientific Model 5681 and 5683
AM1950 and AM1960 Series—Standard platinum resistance thermometer SPRT probes: SPRT's are used to interpolate temperature in the range from -189.3442°C to 660.323°C on the International Temperature Scale of 1990 (ITS-90). They are widely used as standard or reference thermometers to calibrate other thermometers and to measure temperature precisely in primary and secondary laboratories. Digi-Sense SPRTs feature a extremely low drift rate, outstanding stability and repeatability. The AM1950 and AM1960 come with a quartz sheath.
Accuracy is dependent on the estimated measurement uncertainty of the calibration service providing the calibration.
Probes may be available in different lengths and terminations than what is shown in the ordering table above: please contact our Application Specialists for special order options.
$5,886.19 - $6,475.55CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Connector Type Max Temperature (° F) Probe Type
Availability Pricing
COMPARE
Spade Lugs 1238 Secondary Reference PRT/RTD
$5,886.19CAD / Each
COMPARE
Spade Lugs 1238 Secondary Reference PRT/RTD
$6,475.55CAD / Each