This product is now discontinued, View Related Products
Diba Omnifit® Flared PTFE Tubing Assemblies with Click-N-Seal® M6 ISO(M) Acetal Fittings
Flared tubing and fitting assemblies offer fast leak-free connections
  • Flared tubing sits tightly against a flat bottom of the port
  • Only the tube material is in the fluid path
  • Select from 1/16" or 1/8" tube OD
Self-torquing Click-N-Seal fittings create a seal without any tools. As the fitting is tightened the resistance of the cap increases until the correct torque is reached. Acetal fittings are compact in design to easily fit in tight spaces.
$85.16 - $109.03CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Length (ft) Tubing OD (in)
Availability Pricing
Diba Click-N-Seal™ Plus Tubing Assembly, M6 ISO(M) with flare, PTFE/Acetal, 1/16" OD x 24" L; 1/ea RK-21943-54 Mfr # DB200-16858
COMPARE
2 1/16
$109.03CAD / Each
Diba Click-N-Seal™ Plus Tubing Assembly, M6 ISO(M) with flare, PTFE/Acetal, 1/8" OD x 24" L; 1/ea RK-21943-56 Mfr # DB200-16861
COMPARE
2 1/8
$95.18CAD / Each
Diba Click-N-Seal™ Plus Tubing Assembly, M6 ISO(M) with flare, PTFE/Acetal, 1/8" OD x 12" L; 1/ea RK-21943-58 Mfr # DB200-16863
COMPARE
1 1/8
$85.16CAD / Each