This product is now discontinued, View Related Products
Desco Ionized Air Blowers
Automatically maintain a balanced ion output
Portable Ionized Air Blower is compact, lightweight, and ideal for field use. Blower features an emitter cassette that slides out for easy cleaning and replacement. The grounded metal case eliminates high-voltage fields for greater safety. Airflow output can be adjusted for greater worker comfort. Visual and audible alarms and "Clean Me" indicator automatically signal when cleaning and maintenance are required.
Benchtop Ionized Air Blower emits ions in a true laminar flow reducing ion recombination and emitter contamination. AC ionizing system neutralizes static charges at a greater distance while the fixed temperature heater increases worker comfort. Gold-plated emitters provide longer service life.
$920.87 - $1,589.79CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Accuracy Power (VAC) Range
Availability Pricing
Desco 60500 Portable Ionized Air Blower, 120 VAC RK-26809-44 Mfr # 60500
COMPARE
120
$1,321.26CAD / Each
Desco 60505 Portable ionized air blower, 240 VAC RK-26809-46 Mfr # 60505
COMPARE
240
$920.87CAD / Each
Desco 19500 Benchtop Ionized Air Blower, 120 VAC RK-26809-50 Mfr # 19500
COMPARE
120
$1,589.79CAD / Each
Desco 19520 Benchtop Ionized Air Blower, 240 VAC RK-26809-52 Mfr # 19520
COMPARE
220
$1,485.91CAD / Each