This product is now discontinued, View Related Products
Desco Heel and Toe Grounders
Great for tasks requiring mobility
Heel and toe grounders help eliminate the possibility of ESD damage. Tear-resistant, dual-layer conductive rubber heel cups and toe bands have nonmarring interiors, and an RTG (resistance-to-ground) rating of 106 to 107 Ω. Hook and stretch loop fastening assemblies and straps provide an excellent fit for a wide range of shoe types.
$26.08 - $30.72CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Accuracy Power (VAC) Range
Availability Pricing
Desco 17200 Heel Grounder with 1 MegaOhm Resistor RK-26809-12 Mfr # 17200
COMPARE
$28.36CAD / Each
Desco 17202 Heel Grounder with 2 MegaOhm Resistor RK-26809-14 Mfr # 17202
COMPARE
$26.08CAD / Each
Desco 17222 Toe Grounder with 1 MegaOhm Resistor RK-26809-18 Mfr # 17222
COMPARE
$30.72CAD / Each