Defelsko PosiTector Advanced Ultrasonic Coating Thickness Gauges for Non-Metals
Quickly measure coating thickness on all non-metal substrates
  • Ready to measure - no calibration adjustment required to measure most coatings
  • Solvent, acid, oil, water and dust resistant - meets or exceeds IP5X
  • Internal memory for 100,000 readings up to 1000 batches to be view or downloaded via USB, WiFi, Bluetooth®
  • Bluetooth® 4.0 technology for data transfer to a mobile device running the PosiTector App or optional portable printer
  • BLE API available for integration into third-party software
  • Statistics mode continuously displays average, standard deviation, and number of readings while measuring
  • High contrast backlit display for bright or dark environments
  • Certificate of Calibration showing traceability to NIST or PTB included (Long Form)
  • Proven non-destructive ultrasonic technique conforms to ASTM6132 and ISO2808
  • Selectable display languages
The advanced ultrasonic coating thickness gauges non-destructively measure the thickness of coatings on non-metal substrates including wood, plastic, composites, and more. They also measure total thickness of a coating system or up to three individual layer thicknesses in a multi-layer system. Also features easy-to-read graphic display readout for clear detailed analysis of the coating system.
$3,786.61CAD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Display Range
Availability Pricing
DeFelsko PosiTector 200B3 Coating Thickness Gauge, Non-Metals, 0.5-40 mils RK-59784-01 Mfr # 200B3-E
COMPARE
Graphic Readout 0.5 to 40 mil
$3,786.61CAD / Each