Davis Instruments Vantage Vue Wireless Weather Station
Legendary reliability and toughness in a station that is easy to install and use
  • 50 on-screen graphs for comparing current and past weather
  • 22 alarms to warn of dangers such as high winds, possible flooding, and more
  • Optional WeatherLink software for extensive weather analysis and data storage
Made in the USA, the Vantage Vue weather station includes a distinctive LCD console
which provides user interface, data display, and calculations and a sleek yet tough outdoor
integrated sensor suite (ISS). The Vantage Vue is radio-compatible with Vantage Pro2™
weather stations so you can mix-and-match most components.
This self-contained, wireless station gives you all the weather data you need. You'll see
current weather conditions, including indoor and outdoor temperature and humidity,
barometric pressure, wind speed and direction, dew point, and rainfall. Plus, the station
provides weather forecast icons, moon phase, sunrise/sunset times, graphing of weather
trends, alarms, and more. The unique Weather Center function provides additional
information on each weather variable, for today and over the last 25 days.
Station features frequency-hopping spread spectrum radio technology, wireless
transmission up to 1000 feet (305 meters), weather updates every 2.5 seconds, a glowin-
the-dark user-friendly keypad, on-screen data graphing, and numerous other useful
features. Order LCD console and ISS separately or in the convenient weather station kit.
3 PRODUCT OPTIONS
Compareitem specifications
Description Max Temperature (Outside) (° F)
Availability Pricing
Davis Instruments Wireless Station; Console/Sensor Suite RK-30001-97 Mfr # 06250
COMPARE
Wireless Station; Console/Sensor Suite 150
$590.46CAD / Each
Davis Instruments Wireless LCD Console/Receiver RK-30001-98 Mfr # 06351
COMPARE
Wireless LCD Console/Receiver 150
$292.84CAD / Each
Davis Instruments 06357 Wireless Integrated Sensor Suite RK-30001-99 Mfr # 06357
COMPARE
Wireless Integrated Sensor Suite 150
$528.21CAD / Each