This product is now discontinued, View Related Products
Davis Instruments® Vantage Pro 2 Wireless Weather Stations
Versatile wireless weather station complete with rain collector, temperature/humidity sensors and anemometer
  • Wireless integrated sensor suite runs on solar power; lithium battery provides backup at night and during cloudy, wintry days
  • Rain bucket is self-emptying for exceptionally accurate readings
  • Temperature and humidity sensors are located inside the radiation protective shield
  • Sophisticated forecasting algorithm takes wind and temperature into account - not just barometric pressure
  • Over 80 graphs and over 70 alarms available on the console
  • Anemometer is detachable for ultimate flexibility - mount with integrated sensor suite or mount separately using the included 40 ft cable
  • Optional tripod makes installation easy - tilts to mount on roof or uneven terrain
This powerful station transmits data up to 1000 ft or even further with an optional wireless repeater. Measured parameters update every 2.5 seconds as compared to 3 to 15 minutes found on other stations.
$1,402.67 - $2,335.32CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description Max Temperature (Outside) (° F)
Availability Pricing
Davis Instruments 6153 Wireless Weather Station with 24-Hour Fan Aspirated Radiation Shield RK-34542-65 Mfr # 6153
COMPARE
Wireless Weather Station with 24-Hour Fan Aspirated Radiation Shield 150
$1,402.67CAD / Each
Davis Instruments 6163 Wireless Weather Station; UV, Solar, Fan RK-34542-66 Mfr # 6163
COMPARE
Wireless Weather Station; UV/Solar/Fan 150
$2,084.85CAD / Each
Davis Instruments 6152 Wireless Weather Station RK-86403-03 Mfr # 6152
COMPARE
Wireless Weather Station 150
$1,444.66CAD / Each
Davis Instruments 6162 Plus Wireless Weather Station; UV, Solar RK-86403-13 Mfr # 6162
COMPARE
Plus Wireless Weather Station; UV/Solar 150
$2,335.32CAD / Each