This product is now discontinued, View Related Products
Cyclone Sample Mills
Belt-drive mill is USDA-recommended for NIR sample preparation
  • Mills provide fast grinding with consistent particle sizes
  • Ideal for low-moisture materials such as pharmaceuticals, plastics, detergents, grains, forage, coal, and more
The grinding method of these mills uses high-speed impeller rotation and air currents to throw particles into and around the grinding ring. The impact and abrasion reduce particle size until the material is small enough to flow through the exit with the air current. The airflow removes virtually all material from the chamber—no cleaning necessary. The flow through design allows continuous feeding—collect sample in collection bottles up to 1000 mL. Use the direct-drive mill for economical, general purpose grinding. Use the direct-drive mill for economical, general-purpose grinding. The belt-drive mill maximizes particle size consistency—ideal for NIR applications.
$7,047.96 - $9,631.55CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Power (VAC)
Availability Pricing
Direct-drive cyclone sample mill; 115 VAC RK-04187-00 Mfr # 3010-014
COMPARE
115
$7,047.96CAD / Each
Belt-Drive Cyclone Sample Mill, Continuous Feed; 115VAC, 50/60Hz RK-04187-10 Mfr # 3010-030
COMPARE
115
$9,616.82CAD / Each
Belt-Drive Cyclone Sample Mill, Continuous Feed; 220VAC, 50/60Hz RK-04187-15 Mfr # 3010-019
COMPARE
220
$9,631.55CAD / Each