CrystalVu Color Luer Fittings
Available in 6 colors for quick and easy identification: berry, rose, yellow, lime, violet, and orange. Made of polyolefin, a transparent polycarbonate alternative containing no bisphenol-A (BPA).
Superior chemical resistance compared to polycarbonate. Sterilize by autoclave, gamma irradiation, or ethylene oxide. Meets USP Class VI. Animal-derived ingredient-free. Packs of 25.
$32.41 - $36.47CAD / Pkg of 25
36 variations of this product are available.
See All Product Options Below
36 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size-Port 2 (in) Connection Type-Port 1 Description
Availability Pricing
CrystalVu luer fitting, male luer lock x 3/32" hose barb, Berry, 25/pk RK-45520-02 Mfr # LMSLN-1.5 CVU-B 25PK
COMPARE
3/32 Male luer lock luer fitting, male luer lock x 3/32" hose barb, Berry, 25/pk
$33.15CAD / Pkg of 25
CrystalVu luer fitting, male luer lock x 1/8" hose barb, Berry, 25/pk RK-45520-03 Mfr # LMSLN-2 CVU-B 25PK
COMPARE
1/8 Male luer lock luer fitting, male luer lock x 1/8" hose barb, Berry, 25/pk
$35.73CAD / Pkg of 25
CrystalVu luer fitting, male luer lock x 5/32" hose barb, Berry, 25/pk RK-45520-04 Mfr # LMSLN-2.5 CVU-B 25PK
COMPARE
5/32 Male luer lock luer fitting, male luer lock x 5/32" hose barb, Berry, 25/pk
$34.26CAD / Pkg of 25
CrystalVu luer fitting, male luer lock x 3/16" hose barb, Berry, 25/pk RK-45520-05 Mfr # LMSLN-3 CVU-B 25PK
COMPARE
3/16 Male luer lock luer fitting, male luer lock x 3/16" hose barb, Berry, 25/pk
$36.47CAD / Pkg of 25
CrystalVu luer fitting, male luer lock x 1/16" hose barb, Rose, 25/pk RK-45520-11 Mfr # LMSLN-1 CVU-R 25PK
COMPARE
1/16 Male luer lock luer fitting, male luer lock x 1/16" hose barb, Rose, 25/pk
$33.15CAD / Pkg of 25
CrystalVu luer fitting, male luer lock x 5/32" hose barb, Rose, 25/pk RK-45520-14 Mfr # LMSLN-2.5 CVU-R 25PK
COMPARE
5/32 Male luer lock luer fitting, male luer lock x 5/32" hose barb, Rose, 25/pk
$34.26CAD / Pkg of 25
CrystalVu luer fitting, male luer lock x 3/16" hose barb, Rose, 25/pk RK-45520-15 Mfr # LMSLN-3 CVU-R 25PK
COMPARE
3/16 Male luer lock luer fitting, male luer lock x 3/16" hose barb, Rose, 25/pk
$36.47CAD / Pkg of 25
CrystalVu luer fitting, male luer lock x 3/16" hose barb, Yellow, 25/pk RK-45520-25 Mfr # LMSLN-3 CVU-Y 25PK
COMPARE
3/16 Male luer lock luer fitting, male luer lock x 3/16" hose barb, Yellow, 25/pk
$34.99CAD / Pkg of 25
CrystalVu luer fitting, male luer lock x 1/4" hose barb, Yellow, 25/pk RK-45520-26 Mfr # LMSLN-4 CVU-Y 25PK
COMPARE
1/4 Male luer lock luer fitting, male luer lock x 1/4" hose barb, Yellow, 25/pk
$35.73CAD / Pkg of 25
CrystalVu luer fitting, male luer lock x 1/8" hose barb, Lime, 25/pk RK-45520-33 Mfr # LMSLN-2 CVU-L 25PK
COMPARE
1/8 Male luer lock luer fitting, male luer lock x 1/8" hose barb, Lime, 25/pk
$35.73CAD / Pkg of 25
CrystalVu luer fitting, male luer lock x 5/32" hose barb, Lime, 25/pk RK-45520-34 Mfr # LMSLN-2.5 CVU-L 25PK
COMPARE
5/32 Male luer lock luer fitting, male luer lock x 5/32" hose barb, Lime, 25/pk
$34.26CAD / Pkg of 25
CrystalVu luer fitting, male luer lock x 3/16" hose barb, Lime, 25/pk RK-45520-35 Mfr # LMSLN-3 CVU-L 25PK
COMPARE
3/16 Male luer lock luer fitting, male luer lock x 3/16" hose barb, Lime, 25/pk
$34.99CAD / Pkg of 25
CrystalVu luer fitting, male luer lock x 3/32" hose barb, Violet, 25/pk RK-45520-42 Mfr # LMSLN-1.5 CVU-V 25PK
COMPARE
3/32 Male luer lock luer fitting, male luer lock x 3/32" hose barb, Violet, 25/pk
$32.41CAD / Pkg of 25
CrystalVu luer fitting, male luer lock x 1/8" hose barb, Violet, 25/pk RK-45520-43 Mfr # LMSLN-2 CVU-V 25PK
COMPARE
1/8 Male luer lock luer fitting, male luer lock x 1/8" hose barb, Violet, 25/pk
$35.73CAD / Pkg of 25
CrystalVu luer fitting, male luer lock x 5/32" hose barb, Violet, 25/pk RK-45520-44 Mfr # LMSLN-2.5 CVU-V 25PK
COMPARE
5/32 Male luer lock luer fitting, male luer lock x 5/32" hose barb, Violet, 25/pk
$34.26CAD / Pkg of 25
CrystalVu luer fitting, male luer lock x 3/16" hose barb, Violet, 25/pk RK-45520-45 Mfr # LMSLN-3 CVU-V 25PK
COMPARE
3/16 Male luer lock luer fitting, male luer lock x 3/16" hose barb, Violet, 25/pk
$34.99CAD / Pkg of 25
CrystalVu luer fitting, male luer lock x 1/16" hose barb, Orange, 25/pk RK-45520-51 Mfr # LMSLN-1 CVU-O 25PK
COMPARE
1/16 Male luer lock luer fitting, male luer lock x 1/16" hose barb, Orange, 25/pk
$32.41CAD / Pkg of 25
CrystalVu luer fitting, male luer lock x 3/32" hose barb, Orange, 25/pk RK-45520-52 Mfr # LMSLN-1.5 CVU-O 25PK
COMPARE
3/32 Male luer lock luer fitting, male luer lock x 3/32" hose barb, Orange, 25/pk
$32.41CAD / Pkg of 25
CrystalVu luer fitting, male luer lock x 1/8" hose barb, Orange, 25/pk RK-45520-53 Mfr # LMSLN-2 CVU-O 25PK
COMPARE
1/8 Male luer lock luer fitting, male luer lock x 1/8" hose barb, Orange, 25/pk
$34.26CAD / Pkg of 25
CrystalVu luer fitting, male luer lock x 1/4" hose barb, Orange, 25/pk RK-45520-56 Mfr # LMSLN-4 CVU-O 25PK
COMPARE
1/4 Male luer lock luer fitting, male luer lock x 1/4" hose barb, Orange, 25/pk
$34.26CAD / Pkg of 25
Showing 1 - 20 of 36