Cole-Parmer® Essentials Cryogenic Safety Apparel Kits
Stay protected and save money with these bundled kits
  • Gloves protect at temperatures as low as –256°F (–160°C) and multilayered insulated construction provides maximum thermal protection while maintaining flexibility and dexterity
  • Apron's 100% waterproof liner prevents spilled fluid and cold equipment from coming in contact with the torso, and the adjustable plastic fasteners at waist and neck snap easily into place
Water-resistant Cryo-Gloves provide protection in closed cryogenic systems with limited risk of exposure to cryogenic fluids. A stay-dry lining keeps you warm and comfortable. Cryo-Apron is rated for use with LN2 vapor phase and liquefied natural gas (LNG). Lightweight insulation is nontoxic and asbestos-free; provides warmth without bulk. Aprons are machine washable/dryable.
Note: Do not immerse apparel in liquid nitrogen.
$517.16 - $623.24CAD / Each
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Size Description
Availability Pricing
COMPARE
Nylon Medium Cryogenic Safety Kit; Medium Gloves and 36\" Long Apron
$517.16CAD / Each
COMPARE
Nylon Medium Cryogenic Safety Kit; Medium Gloves and 42\" Long Apron
$549.57CAD / Each
COMPARE
Nylon Medium Cryogenic Safety Kit; Medium Gloves and 48\" Long Apron
$581.99CAD / Each
COMPARE
Nylon Large Cryogenic Safety Kit; Large Gloves and 36\" Long Apron
$521.58CAD / Each
COMPARE
Nylon Large Cryogenic Safety Kit; Large Gloves and 42\" Long Apron
$549.57CAD / Each
COMPARE
Nylon Large Cryogenic Safety Kit; Large Gloves and 48\" Long Apron
$581.99CAD / Each
COMPARE
Nylon X-Large Cryogenic Safety Kit; X-Large Gloves and 36\" Long Apron
$518.63CAD / Each
COMPARE
Nylon X-Large Cryogenic Safety Kit; X-Large Gloves and 42\" Long Apron
$549.57CAD / Each
COMPARE
Nylon X-Large Cryogenic Safety Kit; X-Large Gloves and 48\" Long Apron
$581.99CAD / Each
COMPARE
Nylon Medium Cryogenic Safety Kit; Medium Gloves, 42\" Long Apron, and Face Shield
$611.46CAD / Each
COMPARE
Nylon Large Cryogenic Safety Kit; Large Gloves, 42\" Long Apron, and Face Shield
$611.46CAD / Each
COMPARE
Nylon Large Cryogenic Safety Kit; Large Gloves, 48\" Long Apron, and Face Shield
$623.24CAD / Each
COMPARE
Nylon X-Large Cryogenic Safety Kit; X-Large Gloves, 36\" Long Apron, and Face Shield
$589.36CAD / Each
COMPARE
Nylon X-Large Cryogenic Safety Kit; X-Large Gloves, 48\" Long Apron, and Face Shield
$623.24CAD / Each
Showing 1- 14 of 14