This product is now discontinued, View Related Products
Corrugated Switchboard Matting
Protect workers from harmful or deadly electrical shock
  • ASTM D178-01 specifications
High-performance, nonconductive vinyl mats insulate workers, protecting them from deadly shocks generated by high-voltage equipment. Mats are tested and proven to have a high dielectric strength; however the maximum recommended voltage exposure is much less.
Corrugated Switchboard Model 702
Corrugated surface offers additional slip resistance
1/4'' thickness meets Type II, Class 2 specs
3/8'' thickness meets Type II, Class 3 specs
Both thicknesses conforms to ANSI/ASTM D178-01 standards
$31.68 - $4,045.93CAD / Foot of 1
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Thickness (in) Width (in)
Availability Pricing
Wearwell 702.14x2cutbk Corrugated Switchboard Matting, 2' x custom cut to 75', black RK-55555-12 Mfr # 702.14x2cutbk
COMPARE
24
$31.68CAD / Foot of 1
Wearwell 702.14x75cutbk Corrugated Switchboard Matting, 2' x 75', Black RK-55555-14 Mfr # 702.14x2x75BK
COMPARE
0.25 24
$1,467.50CAD / Roll of 1
Wearwell 702.14x3cutbk Corrugated Switchboard Matting, 3' x custom cut to 75', black RK-55555-16 Mfr # 702.14x3cutbk
COMPARE
36
$48.62CAD / Foot of 1
Wearwell Corrugated Switchboard Matting, 3' x 75', black RK-55555-18 Mfr # 702.14x3x75BK
COMPARE
0.25 36
$1,912.46CAD / Roll of 1
Wearwell 702.14x4cutbk Corrugated Switchboard Matting, 4' x custom cut to 75', black RK-55555-20 Mfr # 702.14x4cutbk
COMPARE
48
$78.09CAD / Foot of 1
Wearwell Corrugated Switchboard Matting, 4' x 75', black RK-55555-22 Mfr # 702.14x4x75BK
COMPARE
0.25 48
$3,603.91CAD / Roll of 1
Wearwell 702.38x3cutbk Corrugated Switchboard Matting, 3' x 75', 3/8'' thick, black RK-55555-24 Mfr # 702.38x3x75BK
COMPARE
0.375 36
$4,045.93CAD / Roll of 1
Showing 1- 7 of 7