Corning® Stripette® Sterile Serological Pipettes, Polystyrene
Bidirectional graduations provide choice of ascending and descending scales
  • Anti-drip tip assures accurate delivery
  • Color-coded magnifier stripes make volume reading and size selection easier
  • Negative graduations allow additional working volume
  • Calibrated "to deliver" for accurate dispensing to ±2%
  • Sterile, individually wrapped, nonpyrogenic, DNase-/RNase-free, and Human DNA-free
The Stripette™ serological pipettes are constructed of polystyrene and feature color-coded vertical stripes which are magnified by internal reagents, making the meniscus easy to read for quick volume identification. Uniform mouthpieces fit all standard pipette fillers. Individually wrapped paper/plastic packaging with "Banana peel" design make pipettes easy to open and remove aseptically with one hand, ideal for sterile tissue culture applications, especially while wearing latex gloves, as the wrap reduces static cling.
$257.84 - $751.43CAD / Case of 1000
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Code Graduations (mL) Max Volume (mL)
Availability Pricing
Corning Stripette™ Serological Pipettes, Polystyrene, Sterile, 1 mL; 1000/Cs RK-13001-20 Mfr # 4485
COMPARE
Yellow 0.01 1
$751.43CAD / Case of 1000
Corning Stripette™ Serological Pipettes, Polystyrene, Sterile, 5 mL; 200/Cs RK-13001-40 Mfr # 4487
COMPARE
Blue 0.1 5
$257.84CAD / Case of 200
Corning Stripette™ Serological Pipettes, Polystyrene, Sterile, 10 mL; 200/Cs RK-13001-50 Mfr # 4488
COMPARE
Orange 0.1 10
$272.58CAD / Case of 200