Corning® Sterile Cell Culture Dishes, Polystyrene
Vented lid design provides consistent gas exchange
  • Optically-clear polystyrene allows for easy cell viewing
  • Stacking beads aid in handling
  • Sterilized by gamma radiation and nonpyrogenic
These sterile cell culture dishes are constructed of durable polystyrene and feature stacking beads for easier handling and to prevent accidental lid lift-off. Choose from a variety of dish shapes and surface treatments.
Dish Shape options include a round and square. Round dishes are the industry standard while square plates allow for easier handling and are ideal for the researcher who wishes to culture large numbers of anchorage dependent cells.
Surface Treatment include non-treated, tissue culture (TC) treated, and ultra-low attachment treated. Non-treated dishes are ideal for applications where cell attachment is not desired. TC-treated dishes allow for optimal cell attachment. Ultra-low attachment surface treatment features a covalently bound hydrogel layer that minimizes cell attachment, protein absorption, and cellular activation.
$419.92 - $1,768.07CAD / Case of 500
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Culture Area (cm²) Surface Treatment Well Shape
Availability Pricing
Corning Cell Culture Dish, Sterile, Polystyrene, Round, TC-Treated, 9 cm2; 500/Cs RK-01835-00 Mfr # 430165
COMPARE
9 Tissue Culture Treated Round
$766.16CAD / Case of 500
Corning Cell Culture Dish, Sterile, Polystyrene, Round, TC-Treated, 21 cm2; 500/Cs RK-01835-01 Mfr # 430166
COMPARE
21 Tissue Culture Treated Round
$839.83CAD / Case of 500
Corning Cell Culture Dish, Sterile, Polystyrene, Round, TC-Treated, 55 cm2; 500/Cs RK-01835-03 Mfr # 430167
COMPARE
55 Tissue Culture Treated Round
$1,768.07CAD / Case of 500
Corning Cell Culture Dish, Sterile, Polystyrene, Round, TC-Treated, 150 cm2; 60/Cs RK-01835-05 Mfr # 430599
COMPARE
148 Tissue Culture Treated Round
$456.75CAD / Case of 60
Corning Cell Culture Dish, Sterile, Polystyrene, Square, TC-Treated, 500 cm2; 16/Cs RK-01835-06 Mfr # 431110
COMPARE
500 Tissue Culture Treated Square
$766.16CAD / Case of 16
Corning Cell Culture Dish, Sterile, Polystyrene, Round, Non-Treated, 21 cm2; 500/Cs RK-01835-08 Mfr # 430589
COMPARE
21 Non-Treated Round
$419.92CAD / Case of 500
Corning Cell Culture Dish, Sterile, Polystyrene, Round, Non-Treated, 55 cm2; 500/Cs RK-01835-09 Mfr # 430591
COMPARE
55 Non-Treated Round
$663.03CAD / Case of 500
Corning Cell Culture Dish, Sterile, Polystyrene, Round, Ultra-Low Attachment Treated, 21 cm2; 20/Cs RK-01835-20 Mfr # 3261
COMPARE
21 Ultra-Low Attachment Treated Round
$648.29CAD / Case of 20
Showing 1- 8 of 8