Corning® Class B Polypropylene Volumetric Flasks
Easily view samples within flask
  • Volumetric flasks meet ISO 1042 Class B requirements
  • Autoclavable
These translucent polypropylene (PP) flasks have an individually calibrated graduation line and are complete with tapered PP stopper. Exposure to temperatures above 140°F (60°C) may affect accuracy.
$50.10 - $138.50CAD / Pkg of 1
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Material Tolerance (mL)
Availability Pricing
Corning Class B Volumetric Flasks, Polypropylene, 10 mL RK-06112-80 Mfr # 5641P-10
COMPARE
10 Polypropylene ±0.08
$50.10CAD / Pkg of 1
Corning Class B Volumetric Flasks, Polypropylene, 25 mL RK-06112-82 Mfr # 5641P-25
COMPARE
25 Polypropylene ±0.08
$61.88CAD / Pkg of 1
Corning Class B Volumetric Flasks, Polypropylene, 50 mL RK-06112-84 Mfr # 5641P-50
COMPARE
50 Polypropylene ±0.12
$67.78CAD / Pkg of 1
Corning Class B Volumetric Flasks, Polypropylene, 100 mL RK-06112-86 Mfr # 5641P-100
COMPARE
100 Polypropylene ±0.20
$72.20CAD / Pkg of 1
Corning Class B Volumetric Flasks, Polypropylene, 250 mL RK-06112-88 Mfr # 5641P-250
COMPARE
250 Polypropylene ±0.30
$88.40CAD / Pkg of 1
Corning Class B Volumetric Flasks, Polypropylene, 500 mL RK-06112-90 Mfr # 5641P-500
COMPARE
500 Polypropylene ±0.50
$104.61CAD / Pkg of 1
Corning 5641P-1L PP Plastic Volumetric Flasks, 1 L, 1/Pk RK-06112-92 Mfr # 5641P-1L
COMPARE
1000 Polypropylene ±0.80
$138.50CAD / Pkg of 1
Showing 1- 7 of 7