Costar® Sterile Multiwell Tissue Culture Plates with Ultra Low Attachment Surface Treatment
Unique hydrogel surface inhibits cell attachment
  • Reduce contamination with non-reversible lids with condensation ring
  • Individual alphanumeric codes for well identification
  • Sterilized by gamma radiation and nonpyrogenic
  • Individually wrapped to eliminate cross contamination
These multiwell tissue culture plates have a unique covalently bonded hydrogel surface that is hydrophilc and neutrally charged. It minimizes cell attachment, protein absorption and enzyme activation. The surface is non-cytotoxic, biologically inert, and nondegradable. Use these products to maintain cells in a suspended, unattached state, prevent stem cells from attachment-mediated differentiation, prevent anchorage-dependent cells from dividing, and reduce binding of attachment and serum proteins to the substrate.
$847.20 - $1,001.91CAD / Case of 24
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Culture Area (cm²) Number of Wells Well Volume (mL)
Availability Pricing
Costar Cell Culture Multiwell Plate with Lid, Sterile, Ultra-Low Attachment Treated, 6-Well, 9.5 cm2; 24/Cs RK-01835-24 Mfr # 3471
COMPARE
9.5 6 16.8
$847.20CAD / Case of 24
Costar Cell Culture Multiwell Plate with Lid, Sterile, Ultra-Low Attachment Treated, 24-Well, 1.9 cm2; 24/Cs RK-01835-26 Mfr # 3473
COMPARE
1.9 24 3.4
$869.30CAD / Case of 24
Costar Cell Culture Multiwell Plate with Lid, Sterile, Ultra-Low Attachment Treated, 96-Well, 0.32 cm2; 24/Cs RK-01835-28 Mfr # 3474
COMPARE
0.32 96 0.36
$1,001.91CAD / Case of 24