This product is now discontinued, View Related Products

Corning Wide-Mouth Milk Dilution Bottles

Wide mouth allows for easy pouring and measuring of liquids
  • Autoclavable
These wide-mouth glass bottles meet the requirements for milk dilution bottles stated in the Standard Methods for the Examination of Dairy Products, published by the American Public Health Association. Graduated bottles have a cut-line graduation mark at 99 ± 1 mL. The wide mouth makes pouring and measuring liquids easier, and simplify the addition of dry solids.

Bottles are autoclavable and phenolic cap with rubber liner withstands wet sterilization up to 121°C. Bottles have a 160 mL capacity and measure 150 mm H x 44 mm W x 44 mm D.
$287.31 - $851.62CAD / Pkg of 12

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Material Outside Diameter (OD) (in)
Availability Pricing
Corning 1368-160 Wide-Mouth Milk Dilution Bottle, Non-Graduated, 160mL; 12/Pk RK-34583-00 Mfr # 1368-160
COMPARE
160 Glass 1 3/4
$287.31CAD / Pkg of 12
Corning 1368-160 Wide-Mouth Milk Dilution Bottle, Non-Graduated, 160mL; 48/CS RK-34583-20 Mfr # 1368-160
COMPARE
160 Glass 1 3/4
$735.22CAD / Case of 48
Corning 1373-160 Wide-Mouth Milk Dilution Bottle, Graduated, 160mL; 12/PK RK-34585-00 Mfr # 1373-160
COMPARE
160 Glass 1 3/4
$324.15CAD / Pkg of 12
Corning 1373-160 Wide-Mouth Milk Dilution Bottle, Graduated, 160mL; 48/CS RK-34585-20 Mfr # 1373-160
COMPARE
160 Glass 1 3/4
$851.62CAD / Case of 48
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat