This product is now discontinued, View Related Products

Corning® Gosselin™ Straw Pipettes

Easily reach deep into narrow bottles, vials, and blender bags
  • Ideal for primary dilution sample testing
  • Disposable polypropylene is sterilized to reduce cross-contamination
  • Wide diameter easily handles viscous liquids such as cream and oils
  • Packed in tamper-evident bags with product traceability
Disposable straw pipettes are ideal for transferring tiny amounts of liquids without the need for an exact volume.

Sterilized PP pipettes are packed individually in a tamper-evident bag, 25 pipettes per resealable bag, 40 bags per box, 12 boxes per case.
$450.49 - $849.41CAD / Case of 12000

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Max Volume (mL) Overall Length (mm) Description
Availability Pricing
Corning Gosselin EPI1B-01 Sterile Straw Pipettes, 1 mL, 190 mm, 25/Bag; 12,000/Cs RK-34551-54 Mfr # EPI1B-01
COMPARE
1 190 Sterile Straw Pipettes, 1 mL, 190 mm, 25/Bag; 12,000/Cs
$849.41CAD / Case of 12000
Corning Gosselin EPI1B-05 Sterile Straw Pipettes, 1 mL, 150 mm, 100/Bag; 12,000/Cs RK-34551-55 Mfr # EPI1B-05
COMPARE
1 150 Sterile Straw Pipettes, 1 mL, 150 mm, 100/Bag; 12,000/Cs
$625.09CAD / Case of 12000
Corning Gosselin EPI24B-01 Sterile Straw Pipettes, 2.2 mL, 240 mm, 25/Bag; 6000/Cs RK-34551-56 Mfr # EPI24B-01
COMPARE
2.2 240 Sterile Straw Pipettes, 2.2 mL, 240 mm, 25/Bag; 6000/Cs
$468.91CAD / Case of 6000
Corning Gosselin EPI24B-02 Sterile Straw Pipettes, 2.2 mL, 240 mm, 50/Bag; 6000/Cs RK-34551-57 Mfr # EPI24B-02
COMPARE
2.2 240 Sterile Straw Pipettes, 2.2 mL, 240 mm, 50/Bag; 6000/Cs
$450.49CAD / Case of 6000
Corning Gosselin EPI5-01 Sterile Straw Pipettes, 5 mL, 187 mm, 10/Bag; 3600/Cs RK-34551-58 Mfr # EPI5-01
COMPARE
5 187 Sterile Straw Pipettes, 5 mL, 187 mm, 10/Bag; 3600/Cs
$625.09CAD / Case of 3600
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat