This product is now discontinued, View Related Products
Corning® Gosselin™ Round Bottles
Ideal lightweight and shockproof bottle for media sampling and storage
  • Wide mouth with red screw cap
  • Shock resistant and leakproof
  • Smart choice for sampling and storing liquid, semi-liquid, and solid media
  • Sterile and food contact compliant
Full range of lightweight and leakproof HDPE storage bottles with an easy-access wide mouth. These bottles are easy to fill and empty out its contents. Bottles are fully assembled with a red screw cap for a leakproof seal. Conveniently sterilized to avoid any cross-contamination before final use.
$356.56 - $679.60CAD / Case of 600
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Cap / Stopper Size Material
Availability Pricing
Corning Gosselin Round Bottle, 50 mL, HDPE, 27 mm red screw cap, sterile; 250/cs RK-34551-98 Mfr # LR50A-18
COMPARE
50 27 mm High Density Polyethyene (HDPE)
$606.67CAD / Case of 600
Corning Gosselin Round Bottle, 100 mL, HDPE, 37 mm red screw cap, sterile; 145/cs RK-34551-99 Mfr # LR100-12
COMPARE
100 37 mm High Density Polyethyene (HDPE)
$679.60CAD / Case of 335
Corning Gosselin Round Bottle, 150 mL, HDPE, 37 mm red screw cap, sterile; 140/cs RK-34552-00 Mfr # LR150A-14
COMPARE
150 37 mm High Density Polyethyene (HDPE)
$665.24CAD / Case of 250
Corning Gosselin Round Bottle, 250 mL, HDPE, 37 mm red screw cap, sterile; 68/cs RK-34552-02 Mfr # LR250-09
COMPARE
250 37 mm High Density Polyethyene (HDPE)
$356.56CAD / Case of 145
Corning Gosselin Round Bottle, 500 mL, HDPE, 58 mm red screw cap, sterile; 825/cs RK-34552-03 Mfr # LR500-04
COMPARE
500 58 mm High Density Polyethyene (HDPE)
$551.42CAD / Case of 140
Corning Gosselin Round Bottle, 1000 mL, HDPE, 58 mm red screw cap, sterile; 68/cs RK-34552-04 Mfr # LR1000-08
COMPARE
1000 58 mm High Density Polyethyene (HDPE)
$423.23CAD / Case of 68