Corning® 96-Well Microplates, Polystyrene
Surfaces with different degrees of hydrophilicity—ideal for solid phase immunoassays (ELISA)
  • Formulated for uniform binding and low background absorption
  • Conforms to standard microplate footprint and dimensions
  • Constructed of durable polystyrene
These 96-well polystyrene microplates meet the SBS standard to ensure compatibility with microplate and automation equipment. Plates are available sterile and non-sterile, with and without lids, and come in a wide range of colors, surface treatments, and well shapes.
Plate Colors: Clear plates have excellent optical properties making it ideal for optical measurements. Black plates have low background fluorescence, minimal light scatter and reduced crosstalk. White plates enhance luminescent signals and have low background luminescence and fluorescence.
Surface Treatment options are the standard tissue culture (TC) surface treatment for optimal cell attachment, not treated/medium binding surface treatment which is hydrophobic in nature and binds biomolecules through passive interactions, and high binding treatment which is capable of binding medium (>10kD) and large biomolecules that possess ionic groups and/or hydrophobic regions.
Well Shape options include flat wells which maximize light transmission, round wells which optimize coating and washing, and V-bottom wells let you completely retrieve well contents.
$383.08 - $1,105.04CAD / Case of 100
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
Compareitem specifications
Plate Color Sterile Surface Treatment
Availability Pricing
Corning 96-Well Microplate with Lid, Polystyrene, Sterile, Clear, Not Treated, Flat Bottom; 100/Cs RK-01728-01 Mfr # 3370
COMPARE
Clear Yes Not Treated
$692.49CAD / Case of 100
Corning 96-Well Microplate without Lid, Polystyrene, Non-Sterile, Clear, Not Treated, V-Bottom; 100/Cs RK-01728-05 Mfr # 3897
COMPARE
Clear No Not Treated
$523.05CAD / Case of 100
Corning 96-Well Microplate with Lid, Polystyrene, Sterile, Clear, Not Treated, Round Bottom; 100/Cs RK-01728-07 Mfr # 3788
COMPARE
Clear Yes Not Treated
$685.13CAD / Case of 100
Corning 96-Well Microplate without Lid, Polystyrene, Non-Sterile, Clear, Not Treated, Round Bottom; 100/Cs RK-01728-20 Mfr # 3797
COMPARE
Clear No Not Treated
$427.28CAD / Case of 100
Corning 96-Well Microplate without Lid, Polystyrene, Non-Sterile, Clear, Not Treated, Flat Bottom; 100/Cs RK-01728-26 Mfr # 9017
COMPARE
Clear No Not Treated
$383.08CAD / Case of 100
Corning 96-Well Microplate without Lid, Polystyrene, Non-Sterile, Clear, High Binding, Flat Bottom; 100/Cs RK-01728-28 Mfr # 9018
COMPARE
Clear No High Binding
$847.20CAD / Case of 100
Corning 96-Well Microplate with Lid, Polystyrene, Sterile, Black, TC-Treated, Flat Bottom; 100/Cs RK-01728-62 Mfr # 3916
COMPARE
Black Yes Tissue Culture
$1,075.57CAD / Case of 100
Corning 96-Well Microplate with Lid, Polystyrene, Sterile, White, TC-Treated, Flat Bottom; 100/Cs RK-01728-63 Mfr # 3917
COMPARE
White Yes Tissue Culture
$1,024.01CAD / Case of 100
Corning 96-Well Microplate without Lid, Polystyrene, Non-Sterile, White, High Binding, Flat Bottom; 100/Cs RK-01728-65 Mfr # 3922
COMPARE
White No High Binding
$957.70CAD / Case of 100
Corning 96-Well Microplate without Lid, Polystyrene, Non-Sterile, Black, Not Treated, Flat Bottom; 100/Cs RK-01728-66 Mfr # 3915
COMPARE
Black No Not Treated
$891.40CAD / Case of 100
Corning 96-Well Microplate without Lid, Polystyrene, Non-Sterile, Black, High Binding, Flat Bottom; 100/Cs RK-01728-67 Mfr # 3925
COMPARE
Black No High Binding
$1,105.04CAD / Case of 100
Showing 1- 11 of 11