Corning® 96-Well Clear Bottom Microplates, Polystyrene
Clear bottom plates optimize optical clarity and facilitate cell detection
  • Flat wells maximize light transmission
  • Opaque walls prevent well-to-well cross-talk
  • Optically clear flat bottom permits direct microscopic viewing
  • Constructed of durable polystyrene
These 96-well polystyrene microplates meet the SBS standard to ensure compatibility with microplate and automation equipment. Clear plate bottoms are 60% thinner than conventional polystyrene microplates, resulting in lower background fluorescence and enabling readings down to 340 nm. Plates are available sterile and non-sterile, with and without lids, and come in a range of colors and surface treatments.
Plate Colors: Black plates have low background fluorescence, minimal light scatter and reduced crosstalk. White plates enhance luminescent signals and have low background luminescence and fluorescence.
Surface Treatment options are the standard tissue culture (TC) surface treatment for optimal cell attachment and not treated/medium binding surface treatment which is hydrophobic in nature and binds biomolecules through passive interactions.
$1,399.72 - $1,650.20CAD / Case of 100
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Plate Color Sterile Surface Treatment
Availability Pricing
Corning 96-Well Clear Bottom Microplate with Lid, Polystyrene, Sterile, Black, TC-Treated, Flat Bottom; 100/Cs RK-01728-41 Mfr # 3904
COMPARE
Black Plate - Clear Bottom Yes Tissue Culture
$1,635.46CAD / Case of 100
Corning 96-Well Clear Bottom Microplate with Lid, Polystyrene, Sterile, White, TC-Treated, Flat Bottom; 100/Cs RK-01728-43 Mfr # 3903
COMPARE
White Plate - Clear Bottom Yes Tissue Culture
$1,650.20CAD / Case of 100
Corning 96-Well Clear Bottom Microplate without Lid, Polystyrene, Sterile, Black, Not Treated, Flat Bottom; 100/Cs RK-01728-44 Mfr # 3631
COMPARE
Black Plate - Clear Bottom No Not Treated
$1,399.72CAD / Case of 100