This product is now discontinued, View Related Products
CoorsTek High-Form Crucibles, Porcelain
Glazed inside and out
  • Autoclavable
  • Porcelain material
High-form crucibles are glazed inside and out except for outside bottom and rim.
$78.09 - $963.60CAD / Pkg of 6
18 variations of this product are available.
See All Product Options Below
18 PRODUCT OPTIONS
Compareitem specifications
Overall Height (mm) Capacity (mL) Top Outside Diameter (mm)
Availability Pricing
CoorsTek 60101 High-Form Crucible, Porcelain; 1.3 mL, 15 mm top OD, 15 mm H; 6/PK RK-17803-00 Mfr # 60101
COMPARE
15 1.3 15
$78.09CAD / Pkg of 6
CoorsTek 66103 High-Form Crucible, Porcelain; 5 mL, 24 mm top OD, 20 mm H; 12/PK RK-17803-02 Mfr # 66103
COMPARE
20 5 24
$160.60CAD / Pkg of 12
CoorsTek 66104 High-Form Crucible, Porcelain; 10 mL, 31 mm top OD, 26 mm H; 12/PK RK-17803-04 Mfr # 66104
COMPARE
26 10 31
$182.70CAD / Pkg of 12
CoorsTek 66105 High-Form Crucible, Porcelain; 12 mL, 35 mm top OD, 28 mm H; 12/PK RK-17803-06 Mfr # 66105
COMPARE
28 12 35
$176.81CAD / Pkg of 12
CoorsTek 66107 High-Form Crucible, Porcelain; 20 mL, 40 mm top OD, 36 mm H; 12/PK RK-17803-08 Mfr # 66107
COMPARE
36 20 40
$271.10CAD / Pkg of 12
CoorsTek 66108 High-Form Crucible, Porcelain; 30 mL, 50 mm top OD, 40 mm H; 12/PK RK-17803-10 Mfr # 66108
COMPARE
40 30 50
$284.36CAD / Pkg of 12
CoorsTek 66109 High-Form Crucible, Porcelain; 80 mL,  60 mm top OD, 49 mm H; 12/PK RK-17803-12 Mfr # 66109
COMPARE
49 80 60
$322.67CAD / Pkg of 12
CoorsTek 66110 High-Form Crucible, Porcelain; 120 mL, 70 mm top OD, 56 mm H; 6/PK RK-17803-14 Mfr # 66110
COMPARE
56 120 70
$197.43CAD / Pkg of 6
CoorsTek 66112 High-Form Crucible, Porcelain; 200 mL, 80 mm top OD, 62 mm H; 6/PK RK-17803-16 Mfr # 66112
COMPARE
62 200 80
$262.26CAD / Pkg of 6
CoorsTek 60101 High-Form Crucible, Porcelain; 1.3 mL, 15 mm top OD, 15 mm H; 24/CS RK-17803-20 Mfr # 60101
COMPARE
15 1.3 15
$293.20CAD / Case of 24
CoorsTek 66103 High-Form Crucible, Porcelain; 5 mL, 24 mm top OD, 20 mm H; 72/CS RK-17803-22 Mfr # 66103
COMPARE
20 5 24
$807.42CAD / Case of 72
CoorsTek 66104 High-Form Crucible, Porcelain; 10 mL, 31 mm top OD, 26 mm H; 72/CS RK-17803-24 Mfr # 66104
COMPARE
26 10 31
$891.40CAD / Case of 72
CoorsTek 66105 High-Form Crucible, Porcelain; 12 mL, 35 mm top OD, 28 mm H; 72/CS RK-17803-26 Mfr # 66105
COMPARE
28 12 35
$963.60CAD / Case of 72
CoorsTek 66107 High-Form Crucible, Porcelain, 20 mL, 40 mm OD, 36 mm H; 36/CS RK-17803-28 Mfr # 66107
COMPARE
36 20 40
$757.32CAD / Case of 36
CoorsTek 66108 High-Form Crucible, Porcelain; 30 mL, 50 mm top OD, 40 mm H; 36/CS RK-17803-30 Mfr # 66108
COMPARE
40 30 50
$798.58CAD / Case of 36
CoorsTek 66109 High-Form Crucible, Porcelain; 80 mL, 60 mm top OD, 49 mm H; 24/CS RK-17803-32 Mfr # 66109
COMPARE
49 80 60
$609.98CAD / Case of 24
CoorsTek 66110 High-Form Crucible, Porcelain; 120 mL, 70 mm top OD, 56 mm H; 24/CS RK-17803-34 Mfr # 66110
COMPARE
56 120 70
$713.12CAD / Case of 24
CoorsTek 66112 High-Form Crucible, Porcelain; 200 mL, 80 mm top OD, 62 mm H; 12/CS RK-17803-36 Mfr # 66112
COMPARE
62 200 80
$502.43CAD / Case of 12
Showing 1- 18 of 18