This product is now discontinued, View Related Products
CoorsTek High-Form Crucible Covers, High Alumina
Manufactured from 99.8% pure aluminum oxide, these crucible covers are highly resistant to chemicals, alkalines, and other fluxes. Crucible cover is inactive in hydrogen and carbonaceous atmospheres. Withstands temperatures to 3182°F (1750°C). Crucibles sold separately. Autoclavable.
$113.45 - $187.86CAD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Overall Height (mm) Capacity (mL)
Availability Pricing
CoorsTek 65605 High-Form Crucible Cover, High Alumina, 10 mL RK-17804-11 Mfr # 65605
COMPARE
31 10
$113.45CAD / Each
CoorsTek 65608 High-Form Crucible Cover, High Alumina, 20 mL RK-17804-13 Mfr # 65608
COMPARE
35 20
$120.38CAD / Each
CoorsTek 65611 High-Form Crucible Cover, High Alumina, 50 mL RK-17804-15 Mfr # 65611
COMPARE
47 50
$137.17CAD / Each
CoorsTek 65613 High-Form Crucible Cover, High Alumina, 100 mL RK-17804-17 Mfr # 65613
COMPARE
61 100
$148.08CAD / Each
CoorsTek 65616 High-Form Crucible Cover, High Alumina, 250 mL RK-17804-19 Mfr # 65616
COMPARE
93 250
$187.86CAD / Each