This product is now discontinued, View Related Products
Oakton® Traceable® One-Shot Conductivity Standard Solutions with Calibration
Each pack supplied with a NIST/ISO 34, ISO 17025/A2LA traceable certificate
  • Get accurate, reliable measurements with included NIST-traceable calibration
NIST-Traceable Calibration Standards are the most accurate available. Accuracy at 25°C is ±0.25 microsiemens for 5 and 10 microsiemen solutions and ±0.25% for other solutions or the uncertainty shown on the certificate, whichever is greater.
These convenient single-use vials eliminate contamination concerns and are ideal for the lab as well as the field. Calibration is performed in the large 3-1/2''H x 1-3/4'' dia, PE vial that accommodates virtually any conductivity probe.
Traceable® products are calibrated to NIST-traceable standards and ready to use as soon as you receive it. Each product goes through an intense vetting process before it can carry the Traceable name, and it includes NIST-traceable certification. This means that a 17025-accredited calibration test lab has confirmed that the product meets or exceeds all stated specifications, saving you both time and money by not having to calibrate separately.
NIST-traceable calibration also helps you meet quality standards and regulatory compliance for ISO, FDA, cGMP, VFC, CAP, CLIA and Joint Commission requirements.
$50.46 - $142.55CAD / Pkg of 6
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Conductivity-to-TDS Calibration Values (ppm 442) Conductivity-to-TDS Calibration Values (ppm NaCl) Conductivity Value (µS)
Availability Pricing
COMPARE
5
$142.55CAD / Pkg of 6
COMPARE
10
$50.46CAD / Pkg of 6
COMPARE
100
$64.46CAD / Pkg of 6
COMPARE
1000
$64.46CAD / Pkg of 6
COMPARE
1413
$64.46CAD / Pkg of 6
COMPARE
10000
$64.46CAD / Pkg of 6
COMPARE
100000
$64.46CAD / Pkg of 6
COMPARE
$52.31CAD / Pkg of 6
COMPARE
150000
$64.83CAD / Pkg of 6
COMPARE
200000
$64.83CAD / Pkg of 6
Showing 1- 10 of 10