This product is now discontinued, View Related Products
PYREX® Kuderna-Danish Concentrator Apparatus without Hooks
Efficiently boil away pestistides for accurate testing
  • Keck clamps instead of spring hooks
  • All columns have venting dimples
Kuderna-Danish Concentrators boil away samples to allow testing for pesticides or other pollutants prior to instrumental analysis. Include three-ball Snyder column with venting dimples and graduated collection tube. Available without springs mounted on collection tube.
$626.19 - $649.76CAD / Case of 1
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Corning 2157-500 Kuderna-Danish Concentrator without springs, 500 mL capacity, 615mm H RK-34530-01 Mfr # 2157-500
COMPARE
Kuderna-Danish Concentrator without springs, 500 mL capacity, 615mm H
$649.76CAD / Case of 1
Corning 2157-1L Kuderna-Danish Concentrator without springs, 1L capacity, 720mm H RK-34530-03 Mfr # 2157-1L
COMPARE
Kuderna-Danish Concentrator without springs, 1L capacity, 720mm H
$626.19CAD / Case of 1