Compression Fitting Inserts for Soft Tubing
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
Inserts fit in the end of the soft tubing to provide the rigidity required for adequate compression and a proper seal.
$10.17 - $75.88CAD / Pkg of 25
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Compression Size / Tube Od (Port 1) (in) Description
Availability Pricing
Parker Hannifin P-P-4 Inserts for soft tubing, white PP, 1/4" OD x 5/32" ID; 10/Pk RK-06391-10 Mfr # P-P-4
COMPARE
1/4 Inserts for soft tubing, white PP, 1/4" OD x 5/32" ID; 10/Pk
$11.49CAD / Pkg of 10
Parker Hannifin P-P-5 Inserts for soft tubing, white PP, 5/16" OD x 3/16" ID; 10/Pk RK-06391-20 Mfr # P-P-5
COMPARE
5/16 Inserts for soft tubing, white PP, 5/16" OD x 3/16" ID; 10/Pk
$11.05CAD / Pkg of 10
Parker Hannifin P-P-6 Inserts for soft tubing, white PP, 3/8" OD x 1/4" ID; 10/Pk RK-06391-30 Mfr # P-P-6
COMPARE
3/8 Inserts for soft tubing, white PP, 3/8" OD x 1/4" ID; 10/Pk
$10.17CAD / Pkg of 10
Parker Hannifin P-P-8 Inserts for soft tubing, white PP, 1/2" OD x 3/8" ID; 10/Pk RK-06391-40 Mfr # P-P-8
COMPARE
1/2 Inserts for soft tubing, white PP, 1/2" OD x 3/8" ID; 10/Pk
$10.61CAD / Pkg of 10
Parker Hannifin P4TS3 Inserts for soft tubing, black PP, 1/4" OD x 1/8" ID; 25/Pk RK-06444-91 Mfr # P4TS3
COMPARE
1/4 Inserts for soft tubing, black PP, 1/4" OD x 1/8" ID; 25/Pk
$34.62CAD / Pkg of 25
Parker Hannifin P6TS4 Inserts for soft tubing, black PP, 3/8" OD x 1/4" ID; 25/Pk RK-06444-92 Mfr # P6TS4
COMPARE
3/8 Inserts for soft tubing, black PP, 3/8" OD x 1/4" ID; 25/Pk
$43.10CAD / Pkg of 25
Parker Hannifin P8TS6 Inserts for soft tubing, black PP, 1/2" OD x 3/8" ID; 25/Pk RK-06444-94 Mfr # P8TS6
COMPARE
1/2 Inserts for soft tubing, black PP, 1/2" OD x 3/8" ID; 25/Pk
$75.88CAD / Pkg of 25
Parker Hannifin P10TS8 Inserts for soft tubing, black PP, 5/8" OD x 1/2" ID; 25/Pk RK-06444-96 Mfr # P10TS8
COMPARE
5/8 Inserts for soft tubing, black PP, 5/8" OD x 1/2" ID; 25/Pk
$60.41CAD / Pkg of 25
Showing 1- 8 of 8