Compression Fitting Ferrules
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
Ferrules are required to create a leak-tight seal.
$9.43 - $19.60CAD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Compression Size / Tube Od (Port 1) (in) Description
Availability Pricing
Parker Hannifin 1 TZ-SS Compression Fitting Ferrule, 316SS, 1/16" OD; 1/EA RK-03302-24 Mfr # 1 TZ-SS
COMPARE
Compression Fitting Ferrule, 316SS, 1/16" OD; 1/EA
$19.60CAD / Each
Parker Hannifin 2 TZ-SS Compression Fitting Ferrule, 316SS, 1/8" OD; 1/EA RK-03302-26 Mfr # 2 TZ-SS
COMPARE
Compression Fitting Ferrule, 316SS, 1/8" OD; 1/EA
$10.61CAD / Each
Parker Hannifin 4 TZ-SS Compression Fitting Ferrule, 316SS, 1/4" OD; 1/EA RK-03302-31 Mfr # 4 TZ-SS
COMPARE
Compression Fitting Ferrule, 316SS, 1/4" OD; 1/EA
$9.43CAD / Each
Parker Hannifin 6 TZ-SS Compression Fitting Ferrule, 316SS, 3/8" OD; 1/EA RK-03302-36 Mfr # 6 TZ-SS
COMPARE
Compression Fitting Ferrule, 316SS, 3/8" OD; 1/EA
$10.46CAD / Each
Parker Hannifin 8 TZ-SS Compression Fitting Ferrule, 316SS, 1/2" OD; 1/EA RK-03302-41 Mfr # 8 TZ-SS
COMPARE
Compression Fitting Ferrule, 316SS, 1/2" OD; 1/EA
$14.59CAD / Each