Parker Hannifin Compressed Air Filters
Remove 99. 99% of oil, water, or solid contaminants as small as 0. 01 μm
  • Available in two flow rates: 6 or 22 scfm
Filters are available in two flow rates: 6 or 22 scfm. A manual drain valve maintains continuous coalescing and draining of liquids. Liquids coalesce from the filter cartridge as rapidly as they enter the filter—filter removes liquids for extended periods without loss of efficiency or flow capac ity. Easily replace exhausted filter tubes without tools. Order replacement filter tubes separately.
$163.55 - $373.14CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Free Air Capacity (CFM)
Availability Pricing
Parker Hannifin 8A02N-OB2-BX In-Line Compressed Air Filter, 6 scfm, 150 psi, 1/4"NPT(F) RK-02917-00 Mfr # 8A02N-0B2-BX
COMPARE
6
$163.55CAD / Each
Parker Hannifin 8B02N-OA2-BX Compressed-Air Filter, 22 cfm, 1/4" NPT(F) RK-02917-10 Mfr # 8B02N-0A2-BX
COMPARE
22
$373.14CAD / Each