This product is now discontinued, View Related Products
Complete Vacuum Filtration Systems
Connect to your house vacuum line or order a system with a pump
  • Various filter sizes and types to suit your needs!
  • Composed of durable and chemical-resistant equipment
We've combined our best-selling filtration products into complete kits for your convenience. All kits include a filtering flask, Buchner funnel, one-hole stopper, vacuum tubing, filter forceps, and filter paper. Some kits even include a vacuum pump.
The polypropylene filtering flask has a 45° angle side-arm to help prevent tipping, and accepts 1/4" to 3/8" ID tubing and a size 8 stopper. Autoclavable.
The silicone stopper is a size eight and has one hole for the insertion of the polypropylene Buchner funnel. The funnel is fritted and accepts 90 mm (9 cm) diameter filter papers. The filter forceps help to gently remove the filter from the funnel after use.
$419.92 - $1,049.05CAD / Each
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Vacuum Filtration Kit without Vacuum Pump; 0.45um Pore Filters
$564.31CAD / Each
COMPARE
Vacuum Filtration Kit without Vacuum Pump; 0.2um Pore Filters
$590.83CAD / Each
COMPARE
Vacuum Filtration Kit without Vacuum Pump; Grade 1 Filters
$419.92CAD / Each
COMPARE
Vacuum Filtration Kit without Vacuum Pump; Grade 3 Filters
$443.49CAD / Each
COMPARE
Vacuum Filtration Kit without Vacuum Pump; Grade GA55 Filters
$568.73CAD / Each
COMPARE
Vacuum Filtration Kit without Vacuum Pump; Grade GD120 Filters
$524.53CAD / Each
COMPARE
Vacuum Filtration Kit without Vacuum Pump; Grade GA100 Filters
$576.10CAD / Each
COMPARE
Vacuum Filtration Kit without Vacuum Pump; Grade QR100 Filters
$1,049.05CAD / Each
COMPARE
Vacuum Filtration Kit with Vacuum Pump; Grade GA55 Filters, 120 VAC
$942.23CAD / Each
COMPARE
Vacuum Filtration Kit with Vacuum Pump; Grade GA55 Filters, 230 VAC
$942.23CAD / Each
COMPARE
Vacuum Filtration Kit with Vacuum Pump; Grade GD120 Filters, 120 VAC
$918.66CAD / Each
COMPARE
Vacuum Filtration Kit with Vacuum Pump; Grade GD120 Filters, 230 VAC
$918.66CAD / Each
COMPARE
Vacuum Filtration Kit with Vacuum Pump; 0.2um Pore Filters, 120 VAC
$973.17CAD / Each
COMPARE
Vacuum Filtration Kit with Vacuum Pump; 0.2um Pore Filters, 230 VAC
$973.17CAD / Each
Showing 1- 14 of 14