This product is now discontinued, View Related Products
Combs for Cole-Parmer Vertical Slab Gel Systems 16170
Combs of PTFE fluoropolymer are precision machined to ensure consistent well width and depth. Combs are easy to remove from gels — leaving smooth, clean sample wells.
$85.46 - $113.45CAD / Each
16 variations of this product are available.
See All Product Options Below
16 PRODUCT OPTIONS
Compareitem specifications
Number of Wells Description
Availability Pricing
COMPARE
10 Comb for Vertical Slab Gel System; 10 Wells, 0.75 mm Thick
$92.46CAD / Each
COMPARE
12 Comb for Vertical Slab Gel System; 12 Wells, 0.75 mm Thick
$92.46CAD / Each
COMPARE
14 Comb for Vertical Slab Gel System; 14 Wells, 0.75 mm Thick
$97.98CAD / Each
COMPARE
20 Comb for Vertical Slab Gel System; 20 Wells, 0.75 mm Thick
$97.98CAD / Each
COMPARE
29 Comb for Vertical Slab Gel System; 29 Wells, 0.75 mm Thick
$109.18CAD / Each
COMPARE
30 Comb for Vertical Slab Gel System; 30 Wells, 0.75 mm Thick
$111.98CAD / Each
COMPARE
10 Comb for Vertical Slab Gel System; 10 Wells, 1.0 mm Thick
$92.46CAD / Each
COMPARE
12 Comb for Vertical Slab Gel System; 12 Wells, 1.0 mm Thick
$92.46CAD / Each
COMPARE
14 Comb for Vertical Slab Gel System; 14 Wells, 1.0 mm Thick
$95.03CAD / Each
COMPARE
20 Comb for Vertical Slab Gel System; 20 Wells, 1.0 mm Thick
$95.77CAD / Each
COMPARE
29 Comb for Vertical Slab Gel System; 29 Wells, 1.0 mm Thick
$109.18CAD / Each
COMPARE
30 Comb for Vertical Slab Gel System; 30 Wells, 1.0 mm Thick
$113.45CAD / Each
COMPARE
10 Comb for Vertical Slab Gel System; 10 Wells, 1.5 mm Thick
$85.46CAD / Each
COMPARE
12 Comb for Vertical Slab Gel System; 12 Wells, 1.5 mm Thick
$92.46CAD / Each
COMPARE
14 Comb for Vertical Slab Gel System; 14 Wells, 1.5 mm Thick
$95.77CAD / Each
COMPARE
20 Comb for Vertical Slab Gel System; 20 Wells, 1.5 mm Thick
$95.77CAD / Each
Showing 1- 16 of 16