Dynalon® Combination Inoculating Needles and Loops
Dual purpose design features an inoculating loop and needle
  • Constructed of flexible polystyrene providing high resistance to breakage
  • Sterile and disposable to reduce risk of cross contamination
These sterile combination inoculating loops and needles have been manufactured to allow flexibility and high resistance to breakage. One end of the inoculating loop has been designed to be used as an inoculating needle, giving these loops a double purpose. Color-coded for easy identification. Case of 1000 (20/bag, 50 bg/cs).
$318.25 - $332.99CAD / Case of 1000
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Color Contained Volume (µL) Sterile
Availability Pricing
Dynalon Combination Inoculating Needle and Loop, Sterile, Polystyrene, 10 µL, 4 mm Loop ID, Blue; 1000/Cs RK-14200-00 Mfr # 226505
COMPARE
Blue 10 Yes
$318.25CAD / Case of 1000
Dynalon Combination Inoculating Needle and Loop, Sterile, Polystyrene, 1 µL, 1 mm Loop ID, Natural; 1000/Cs RK-14200-01 Mfr # 226515
COMPARE
Natural 1 Yes
$332.99CAD / Case of 1000