Idex® Column Couplers for Chromatography
Great for reducing overall flow path volume
  • Couple two devices together
  • Lessen volume of flow path
  • 10-32 coned port connections
Column Couplers allow zero dead-volume connections between column and column, column and detector, or column and guard column.
$38.01 - $100.49CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Idex Column Coupler; PCTFE, 10-32 Coned; 1/EA RK-01939-95 Mfr # U-283
COMPARE
Column Coupler, PCTFE, 10-32 Coned; 1/EA
$78.68CAD / Each
Idex Column Coupler, Acetal/PEEK, 10-32 Coned; 1/EA RK-01939-96 Mfr # U-284
COMPARE
Column Coupler, Acetal/PEEK, 10-32 Coned; 1/EA
$38.01CAD / Each
Idex Coupler, PEEK, Male/Male-1/2" RK-42719-40 Mfr # U-288
COMPARE
Column Coupler, PEEK, Male/Male-1/2,\" 10-32 Coned; 1/EA
$100.49CAD / Each