This product is now discontinued, View Related Products
Cole-Parmer Weighing Funnels, Glass
Clear borosilicate glass funnels are perfect for weighing and transporting powder
  • Wide range of sizes available to suit both general and analytical laboratory needs
  • 1-3/8”(35 mm) long tubular arm allows a quick, mess-free transfer of contents into a volumetric flask
  • One flat side for stability — will not roll off your weighing pan
  • Maximum temperature of 460°F (240°C) for normal use and 750°F (400°C) for short-term use
Autoclavable
$67.78 - $83.39CAD / Pkg of 6
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Stem OD (mm) Stopper Number
Availability Pricing
COMPARE
3 8 9
$67.78CAD / Pkg of 6
COMPARE
6 12 13
$69.25CAD / Pkg of 6
COMPARE
10 15 16
$75.14CAD / Pkg of 6
COMPARE
20 15 16
$72.93CAD / Pkg of 6
COMPARE
40 19 20
$83.39CAD / Pkg of 6