Cole-Parmer Vortex Mixer
True circular orbit for efficient mixing
  • Variable speed is useful for a variety of applications from general mixing to resuspending pellets
Shaker features two modes for both "touch" and continuous operation. The standard "Cup" head supplied with the vortexer will accommodate a single test tube. The head is easily switched with optional heads (sold separately) that accommodate tubes of a variety of sizes as well as microplates. No tools required. It is lightweight and stable during operation due to the optimized counter balance. Safe for cold room or incubator use.
$382.34 - $388.61CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC)
Availability Pricing
COMPARE
120
$382.34CAD / Each
COMPARE
230
$388.61CAD / Each