This product is now discontinued, View Related Products
Cole-Parmer® Class A Glass Volumetric Flasks
Clear borosilicate glass volumetric flasks with ground-glass pennyhead stoppers
  • Blue markings for spot and capacity, and white marking for level
  • Volumetric flasks are calibrated "To Contain" (TC) by ISO, DIN and ASTM standards at 20°C
  • Autoclavable
These Cole-Parmer® volumetric flasks feature Class A accuracy exceeds ASTM E288 and DIN 12664 specifications. These flasks can withstand a maximum temperature of 460°F (240°C) for normal use, and 750°F (400°C) for short term use.
$77.43 - $401.79CAD / Pkg of 6
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Stopper Number Accepted Tolerance (mL)
Availability Pricing
COMPARE
100 ±0.10
$226.31CAD / Pkg of 6
COMPARE
200 ±0.15
$285.84CAD / Pkg of 6
COMPARE
250 ±0.12
$305.14CAD / Pkg of 6
COMPARE
500 ±0.20
$401.79CAD / Pkg of 6
COMPARE
1000 ±0.3
$77.43CAD / Pkg of 1
COMPARE
2000 ±0.5
$97.91CAD / Pkg of 1