Cole-Parmer® VP-200 Series Air Cadet® Vacuum/Pressure Pumps
Unique cavity configuration minimizes dead space and air entrapment
  • No maintenance needed—motors are brushless with sealed ball bearings
  • Worry-free pumping with reduced stress, wear, and heat buildup
Single-head vacuum pumps are economically priced and can handle many tough vacuum/pressure applications. Pumps feature an all-plastic, convoluted diaphragm design constructed with a Noryl™ pump head, FKM diaphragm, PTFE valves, and polyethylene (PE) adapters.
NOTE: Do not use metal fittings with any of these pumps.
$674.81 - $1,312.79CAD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Free-Air Capacity (CFM) Max Vacuum (in Hg) Power (VAC)
Availability Pricing
COMPARE
0.61 20
$759.53CAD / Each
COMPARE
0.61 20 115
$674.81CAD / Each
COMPARE
0.52 20 230
$695.44CAD / Each
COMPARE
0.61 20 115
$1,282.59CAD / Each
COMPARE
0.52 20 230
$1,312.79CAD / Each