This product is now discontinued, View Related Products
Cole-Parmer Ultraviolet/White Light Transilluminators
Side-by-side white light and UV sources save space and money
  • Hinged UV-blocking cover adjusts to suit the user’s viewing angle and to provide easy access to the surface
  • UV side utilizes a long-life filter
The transilluminators provide both a white light area and UV area, each measuring 20 x 20 cm. Easily switch between light sources with rocker switch. View Coomassie Blue stained gels, silver-stained protein gels, autoradiographs, and microtiter plates on the white light side. View ethidium bromide gels on the UV side. Transilluminators feature 5000 hour long-life UV filters. Durable, scratch-resistant epoxy-powder paint finish is easy to clean and maintain. Cover blocks 99% of harmful UV radiation.
$2,909.95 - $4,471.74CAD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Power (Hz) Power (VAC) Description
Availability Pricing
COMPARE
60 115 302nm Wavelength UV & White Light Transilluminator, 115VAC
$3,743.88CAD / Each
COMPARE
50 230 302nm Wavelength UV & White Light Transilluminator, 230VAC
$4,471.74CAD / Each
COMPARE
60 115 365nm Wavelength UV & White Light Transilluminator, 115VAC
$2,909.95CAD / Each
COMPARE
50 230 365nm Wavelength UV & White Light Transilluminator, 230VAC
$3,572.97CAD / Each
COMPARE
60 115 302nm/365nm Wavelength UV & White Light Transilluminator, 115VAC
$4,103.39CAD / Each
COMPARE
50 230 302nm/365nm Wavelength UV & White Light Transilluminator, 230VAC
$4,206.53CAD / Each